Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminopenicillanic acid, 96%

Catalog No. AC104250250
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle

Catalog No. AC104250250

Supplier: Thermo Scientific Chemicals 104250250

Only null left
Add to Cart

CAS: 551-16-6 | C8H12N2O3S | 216.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-16-6
Molecular Formula C8H12N2O3S
Molecular Weight (g/mol) 216.26
MDL Number MFCD00005176
InChI Key NGHVIOIJCVXTGV-ALEPSDHESA-N
Synonym 6-aminopenicillanic acid, 6-apa, penicin, penin, aminopenicillanic acid, 6-aps, phenacyl 6-aminopenicillinate, 6-aminopenicillamine acid, 6-aminopenicillanate, 2s,5r,6r-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid
PubChem CID 11082
ChEBI CHEBI:16705
IUPAC Name (2S,5R,6R)-6-amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 197°C to 203°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 454
Specific Rotation + 276.3
Formula Weight 216.25
Percent Purity 96%
Physical Form Fine Powder
Chemical Name or Material 6-Aminopenicillanic acid, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-993-4
RTECSNumber XH8225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.