Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminouracil 98.0+%, TCI America™
SDP

Catalog No. A0658250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0658250G 250 g
A065825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0658250G Supplier TCI America Supplier No. A0658250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 873-83-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006071
InChI Key LNDZXOWGUAIUBG-UHFFFAOYSA-N
Synonym 6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
PubChem CID 70120
IUPAC Name 6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES NC1=CC(=O)NC(=O)N1

Specifications

Melting Point 360°C
Color White-Yellow
Quantity 250 g
Formula Weight 127.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminouracil
EINECSNumber (9)-0065
RTECSNumber YQ8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.