Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aza-2-thiothymine, 98%

Catalog No. aaa1416706
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAA1416706

Supplier: Thermo Scientific Chemicals A1416706

Only null left
Add to Cart

CAS: 615-76-9 | C4H5N3OS | 143.164 g/mol

6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

Solubility
Soluble in water (partly), and formic acid (5 %).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-76-9
Molecular Formula C4H5N3OS
Molecular Weight (g/mol) 143.164
MDL Number MFCD00006458
InChI Key NKOPQOSBROLOFP-UHFFFAOYSA-N
Synonym 6-aza-2-thiothymine, 2-thio-6-azathymine, 6-azathiothymine, 6-methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5 2h-one, 3-mercapto-6-methyl-1,2,4-triazin-5 2h-one, 1,2,4-triazin-5 2h-one, 3,4-dihydro-6-methyl-3-thioxo, 3-mercapto-6-methyl-as-triazin-5-ol, as-triazin-5-ol, 3-mercapto-6-methyl, 6-methyl-3-sulfanyl-1,2,4-triazin-5-ol, 3,4-dihydro-6-methyl-3-thioxo-1,2,4-triazin-5 2h-one
PubChem CID 1263666
IUPAC Name 6-methyl-3-sulfanylidene-2H-1,2,4-triazin-5-one
SMILES CC1=NNC(=S)NC1=O

Specifications

Melting Point 218°C to 221°C
Quantity 5 g
Beilstein 4861
Solubility Information Soluble in water (partly),and formic acid (5 %).
Formula Weight 143.17
Percent Purity 98%
Chemical Name or Material 6-Aza-2-thiothymine
EINECSNumber 210-445-4
RTECSNumber XZ2853000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.