Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Azaindole, 97+%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
6-Azaindole is an indole derivative and useful synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Azaindole is an indole derivative and useful synthetic intermediate.
Solubility
Soluble in water (hardly), methanol and chloroform.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.
6-Azaindole is an indole derivative and useful synthetic intermediate.
Solubility
Soluble in water (hardly), methanol and chloroform.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.

Specifications
Specifications
Quantity | 5 g |
Beilstein | 109685 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in water (hardly),methanol and chloroform. |
Formula Weight | 118.14 |
Percent Purity | ≥98% |
Physical Form | Solid |
Chemical Name or Material | 6-Azaindole |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
RTECSNumber | UY8720000 |
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction