Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Azaindole, ≥98%

Click to view available options
Quantity:
250mg
1g
5g
271-29-4
C7H6N2
118.139
MFCD01686899
XLKDJOPOOHHZAN-UHFFFAOYSA-N
6-azaindole,1h-pyrrolo 2,3-c pyridine,harmyrine,1,6-diazaindene,1h-pyrrolo 2,3-c pyridine,6h-pyrrolo 2,3-c pyridine,6-azaindol,pyrrolo 2,3-c pyridine,6-aza-6h-indole,zlchem 439
9219
1H-pyrrolo[2,3-c]pyridine
C1=CNC2=C1C=CN=C2
271-29-4
C7H6N2
118.139
MFCD01686899
XLKDJOPOOHHZAN-UHFFFAOYSA-N
6-azaindole,1h-pyrrolo 2,3-c pyridine,harmyrine,1,6-diazaindene,1h-pyrrolo 2,3-c pyridine,6h-pyrrolo 2,3-c pyridine,6-azaindol,pyrrolo 2,3-c pyridine,6-aza-6h-indole,zlchem 439
9219
1H-pyrrolo[2,3-c]pyridine
C1=CNC2=C1C=CN=C2
Applications
6-Azaindole is an indole derivative and useful synthetic intermediate.

Solubility
Soluble in water (hardly), methanol and chloroform.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.
Sensitivity Air & Light Sensitive
Quantity 1g
Beilstein 109685
Formula Weight 118.14
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material 6-Azaindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UY8720000

storageNote1 : 2° to 8°C