Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Azathymine 98.0+%, TCI America™
SDP

Catalog No. a05561g
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 932-53-6
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006457,MFCD00127802
InChI Key XZWMZFQOHTWGQE-UHFFFAOYSA-N
Synonym 6-azathymine, 6-methyl-1,2,4-triazine-3,5 2h,4h-dione, 5-methyl-6-azauracil, azathymine, usaf cb-28, 1,2,4-triazine-3,5 2h,4h-dione, 6-methyl, 6-methyl-1,2,4-triazine-3,5-diol, 6-methyl-1,2,4-triazine-3,5-dione, unii-zwx58t58za, azathymine, 6
PubChem CID 70269
IUPAC Name 6-methyl-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
SMILES CC1=NNC(=O)NC1=O

Specifications

Melting Point 214°C
Color White
Quantity 1 g
Formula Weight 127.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Azathymine
RTECSNumber XY8050000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.