Learn More
6-Azauridine
Description
6-Azauridine can be applied in antiviral studies. 6-azauridine inhibits de novo pyrimidine synthesis and DNA synthesis and is converted intracellularly into mono, di, and triphosphate derivatives, which incorporate into RNA and inhibit protein synthesis. It is also used as an antineoplastic antimetabolite. It interferes with pyrimidine biosynthesis thereby preventing formation of cellular nucleic acids. As the triacetate, it is also effective as an antipsoriatic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Azauridine can be applied in antiviral studies. 6-azauridine inhibits de novo pyrimidine synthesis and DNA synthesis and is converted intracellularly into mono, di, and triphosphate derivatives, which incorporate into RNA and inhibit protein synthesis. It is also used as an antineoplastic antimetabolite. It interferes with pyrimidine biosynthesis thereby preventing formation of cellular nucleic acids. As the triacetate, it is also effective as an antipsoriatic.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C

Specifications
Specifications
Melting Point | 160°C to 161°C |
Color | White |
Quantity | 1 g |
Merck Index | 14,904 |
Solubility Information | It is soluble in water. |
Formula Weight | 245.19 |
Physical Form | Powder |
Chemical Name or Material | 6-Azauridine |
Safety and Handling
Hazard Category | H302+H312+H332-H351 |
Hazard Statement | GHS H Statement H351-H302-H312-H332 Suspected of causing cancer. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
EINECSNumber | 200-199-6 |
RTECSNumber | XY8575000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.