Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Benzyladenine, 99%

Catalog No. p-7022074
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 1214-39-7 | C12H11N5 | 225.26 g/mol

Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.

Notes
Incompatible with strong oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

Melting Point 229°C to 233°C
Assay Percent Range 99%
Quantity 1 g
Beilstein 19406
Solubility Information Soluble in water,methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Formula Weight 225.26
Percent Purity 99%
Chemical Name or Material 6-Benzyladenine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-927-5
RTECSNumber AU6252200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.