6-Benzyladenine, 99%

Manufacturer: Alfa Aesar A1467803

Quantity 1g

 View more versions of this product

Catalog No. AAA1467803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1214-39-7
    Molecular Formula C12H11N5
    Formula Weight 225.26
    MDL Number MFCD00005572
    Beilstein 19406
    Chemical Name or Material 6-Benzyladenine

    Specifications

    Formula Weight 225.26
    Quantity 1g
    TSCA TSCA
    MDL Number MFCD00005572
    Beilstein 19406
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose o
    RTECS Number AU6252200
    Assay Percent Range 99%
    Molecular Formula C12H11N5
    CAS 1214-39-7
    Chemical Name or Material 6-Benzyladenine
    Assay 99%

    Description

    Applications
    Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

    Solubility
    Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.

    Notes
    Incompatible with strong oxidizing agent.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose o

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit