Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%

Catalog No. AAH27463MD
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH27463MD

Supplier: Thermo Scientific Chemicals H27463MD

Only null left
Add to Cart

CAS: 1073354-27-4 | C18H23BN2O2 | 310.204 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-27-4
Molecular Formula C18H23BN2O2
Molecular Weight (g/mol) 310.204
MDL Number MFCD06798270
InChI Key BWIHFPRFKBKWMU-UHFFFAOYSA-N
Synonym 6-benzylamino pyridine-3-boronic acid pinacol ester, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, n-benzyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, amtb753, 6-benzylamino pyridine-3-boronicacidpinacolester, 2-benzylamino pyridine-5-boronic acid pinacol ester, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 44755164
IUPAC Name N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NCC3=CC=CC=C3

Specifications

Melting Point 135°C to 138°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 310.2
Percent Purity 95%
Chemical Name or Material 6-(Benzylamino)pyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.