Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 6-Benzylaminopurine Monoclonal Antibody (3D3)
GREENER_CHOICE

Catalog No. PIMA549741
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549741 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549741 Supplier Invitrogen™ Supplier No. MA549741
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

6-Benzylaminopurine (6-BAP) is a synthetic cytokinin plant growth regulator that is commonly used in plant tissue culture and agricultural practices. It acts as a hormone to promote cell division and growth, leading to increased yield and quality of crops. 6-BAP has been shown to improve the rooting and shoot formation of plant cuttings, as well as increase fruit set and size in various crops. It is also used in the production of ornamental plants and for the rejuvenation of aging plants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 6-Benzylaminopurine
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3D3
Concentration 17.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of 6-BA hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.