Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Benzylaminopurine, MP Biomedicals

Catalog No. mp021009125 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mg

  • 6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin which together with auxins elicits plant growth and development responses
  • BAP is a widely use cytokinin supplement to plant growth media such as Murashige and Skoog medium, Gamborg’s medium, and Chu’s N6 medium
  • Inhibitor of respiratory kinase in plants
  • Increases post-harvest life of green vegetables

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

Melting Point 226°C to 236°C
Color White
Linear Formula C12H11N5
Quantity 500 mg
Solubility Information Soluble in glacial Acetic Acid (50mg/mL). Soluble in concentrated ammonium hydroxide, 0.1 N Sodium hydroxide (10mg/mL - slightly hazy and yellowish solution), methanol, or dioxane.
Formula Weight 225.25
Physical Form Powder
Chemical Name or Material 6-BENZYLAMINOPURINE
Recommended Storage Store at Room Temperature (15-30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.