Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Benzyloxyindole 98.0+%, TCI America™
SDP

Catalog No. B280925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B280925G 25 g
B28091G 1 g
B28095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B280925G Supplier TCI America Supplier No. B280925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15903-94-3
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
InChI Key FPMICYBCFBLGOZ-UHFFFAOYSA-N
Synonym 6-benzyloxyindole, 6-benzyloxy-1h-indole, 1h-indole, 6-phenylmethoxy, 6-phenylmethoxy indole, 6-benzoxyindole, 6-benzyloxy-indole, 6-benzyloxy indole, 6-phenylmethoxyindole, pubchem7315
PubChem CID 260804
IUPAC Name 6-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)C=CN3

Specifications

Melting Point 117°C
Quantity 25 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Benzyloxyindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.