Alfa Aesar™ 6-Bromo-1-indanone, 97%

Manufacturer: Alfa Aesar™ H5400306

Quantity 5g

 View more versions of this product

Catalog No. AAH5400306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14548-39-1
    Molecular Formula C9H7BrO
    Formula Weight 211.06
    MDL Number MFCD02179286
    Chemical Name or Material 6-Bromo-1-indanone

    Specifications

    Formula Weight 211.06
    Melting Point 110°C
    Quantity 5g
    Molecular Formula C9H7BrO
    MDL Number MFCD02179286
    CAS 14548-39-1
    Chemical Name or Material 6-Bromo-1-indanone
    Assay 97%

    Description

    Applications
    6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit