Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-1-indanone, 97%

Catalog No. p-7035795
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH5400303

Supplier: Thermo Scientific Chemicals H5400303

Only null left
Add to Cart

CAS: 14548-39-1 | C9H7BrO | 211.058 g/mol

6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14548-39-1
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD02179286
InChI Key SEQHEDQNODAFIU-UHFFFAOYSA-N
Synonym 6-bromo-1-indanone, 6-bromoindanone, 6-bromo-2,3-dihydro-1h-inden-1-one, 6-bromo-indan-1-one, 6-bromoindan-1-one, 1-indanone, 6-bromo, 1h-inden-1-one, 6-bromo-2,3-dihydro, 6-bromo indanone, 6-bromo-indanone
PubChem CID 139778
IUPAC Name 6-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=CC(=C2)Br

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-indanone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.