Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-chloroquinoline, 96%, Thermo Scientific Chemicals

Catalog Number AAH5489503
Click to view available options
Quantity:
1 g
5 g
1810-71-5
C9H5BrClN
242.50
MFCD04115272
YXRDWUJAJLDABJ-UHFFFAOYSA-N
12894086
6-bromo-2-chloroquinoline
ClC1=NC2=CC=C(Br)C=C2C=C1
1810-71-5
C9H5BrClN
242.50
MFCD04115272
YXRDWUJAJLDABJ-UHFFFAOYSA-N
12894086
6-bromo-2-chloroquinoline
ClC1=NC2=CC=C(Br)C=C2C=C1

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Methanol

Notes
Store at room temperature with proper ventilation.
TRUSTED_SUSTAINABILITY
Melting Point 148°C to 150°C
Quantity 1 g
Synonym 6-bromo-2-chloro-quinoline, 2-chloro-6-bromoquinoline, quinoline, 6-bromo-2-chloro, pubchem14631, acmc-209ehe, 6-bromo-2 chloroquinoline, 2-chloro-6-bromo-quinoline, ksc537o9r, quinoline,6-bromo-2-chloro, abbypharma ap-14-10831
Solubility Information Soluble in Chloroform and Methanol
Formula Weight 242.5
Percent Purity 96%
Chemical Name or Material 6-Bromo-2-chloroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit