Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-3-(3-oxycyclopentyl)indole, 95%, Thermo Scientific™

Catalog No. AAH6385803
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676170-06-2
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.149
InChI Key TWDVYIPKPMKBCL-UHFFFAOYSA-N
Synonym 3-6-bromo-1h-indol-3-yl cyclopentanone, 3-6-bromo-1h-indol-3-yl cyclopentan-1-one
PubChem CID 22139468
IUPAC Name 3-(6-bromo-1H-indol-3-yl)cyclopentan-1-one
SMILES C1CC(=O)CC1C2=CNC3=C2C=CC(=C3)Br

Specifications

Quantity 1 g
Beilstein 21019370
Formula Weight 278.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Bromo-3-(3-oxycyclopentyl)indole
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.