Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromopyridine-3-boronic acid, 95%

Catalog No. AAL2008506
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2008506 5 g
AAL20085MD 250 mg
AAL2008503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2008506 Supplier Thermo Scientific Chemicals Supplier No. L2008506
Add to Cart

CAS: 223463-14-7 | C5H5BBrNO2 | 201.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223463-14-7
Molecular Formula C5H5BBrNO2
Molecular Weight (g/mol) 201.81
MDL Number MFCD03411558
InChI Key BCYWDUVHAPHGIP-UHFFFAOYSA-N
Synonym 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid
PubChem CID 2762706
IUPAC Name (6-bromopyridin-3-yl)boronic acid
SMILES OB(O)C1=CN=C(Br)C=C1

Specifications

Melting Point ∽165°C (decomposition)
Quantity 5 g
Beilstein 9119918
Formula Weight 201.82
Percent Purity 95%
Chemical Name or Material 6-Bromopyridine-3-boronic acid
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.