Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-1-hexyne, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
10297-06-0
C6H9Cl
116.59
MFCD00013697
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chloro-1-hexyne,5-hexynechloride,1-hexyne,6-chloro,1-hexyne, 6-chloro,5-hexynyl chloride,1-chloro-5-hexyne,6-chloro-hex-1-yne,acmc-1boe0,zukocgmvjuxija-uhfffaoysa
272001
6-chlorohex-1-yne
C#CCCCCCl
10297-06-0
C6H9Cl
116.59
MFCD00013697
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chloro-1-hexyne,5-hexynechloride,1-hexyne,6-chloro,1-hexyne, 6-chloro,5-hexynyl chloride,1-chloro-5-hexyne,6-chloro-hex-1-yne,acmc-1boe0,zukocgmvjuxija-uhfffaoysa
272001
6-chlorohex-1-yne
C#CCCCCCl
Density 0.9620g/mL
Refractive Index 1.4490 to 1.451
Boiling Point 131°C
Flash Point 37°C
Specific Gravity 0.962
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH∽C(CH2)4Cl
Formula Weight 116.59
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-hexyne
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning