Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals

Catalog No. AAH25825ME
Click to view available options
Quantity:
100 mg
500 mg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1651-29-2
Molecular Formula C5H2ClFN4
Molecular Weight (g/mol) 172.55
MDL Number MFCD02183557
InChI Key UNRIYCIDCQDGQE-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoropurine, 6-chloro-2-fluoro-9h-purine, 2-fluoro-6-chloropurine, 6-chloro-2-fluoro-purine, cfp, 1jdj, pubchem9261, acmc-1c2uh, 2-fluoro,6-chloro-purine, 2-fluoro-6-chloro-purine
PubChem CID 5287914
IUPAC Name 6-chloro-2-fluoro-7H-purine
SMILES FC1=NC(Cl)=C2NC=NC2=N1

Specifications

Melting Point 151°C to 153°C
Quantity 500 mg
Formula Weight 172.55
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoropurine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.