Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-2-picoline, 98%

Catalog No. AC152230500
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle

CAS: 18368-63-3 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18368-63-3
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00006245
InChI Key GXZDYRYYNXYPMQ-UHFFFAOYSA-N
Synonym 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
PubChem CID 87601
IUPAC Name 2-chloro-6-methylpyridine
SMILES CC1=CC=CC(Cl)=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 64°C to 68°C (10.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5260 to 1.528
Linear Formula CH3(C5H3N)Cl
Quantity 50 g
Beilstein 20, I, 84
Specific Gravity 1.16
Formula Weight 127.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 6-chloro-2-picoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 242-241-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.