Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Chloro-3-indolyl beta-D-Galactopyranoside 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
20 mg
100 mg
Specifications
Specifications
Color | White-Yellow |
Quantity | 100 mg |
Formula Weight | 329.73 |
Percent Purity | ≥98.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 6-Chloro-3-indolyl beta-D-Galactopyranoside [for Biochemical Research] |
Safety and Handling
TSCA | No |
Recommended Storage | Freezer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction