Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-4-chromanone 98.0+%, TCI America™
SDP

Catalog No. C196825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C196825G 25 g
C19685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C196825G Supplier TCI America Supplier No. C196825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37674-72-9
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
InChI Key LLTDYHFVIVSQPJ-UHFFFAOYSA-N
Synonym 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5
PubChem CID 95273
IUPAC Name 6-chloro-2,3-dihydrochromen-4-one
SMILES C1COC2=C(C1=O)C=C(C=C2)Cl

Specifications

Melting Point 101°C
Quantity 25 g
Formula Weight 182.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-4-chromanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.