Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-4-quinolinol 98.0+%, TCI America™
SDP

Catalog No. C33121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33121G 1 g
C33125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33121G Supplier TCI America Supplier No. C33121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23432-43-1
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00024011
InChI Key XXGUQCVVGPZTPF-UHFFFAOYSA-N
Synonym 6-Chloro-4-hydroxyquinoline
PubChem CID 220929
IUPAC Name 6-chloro-1H-quinolin-4-one
SMILES C1=CC2=C(C=C1Cl)C(=O)C=CN2

Specifications

Melting Point 269°C
Quantity 1 g
Formula Weight 179.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-4-quinolinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.