Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-5-(2-chloroethyl)oxindole 98.0+%, TCI America™
SDP

Catalog No. C26515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26515G 5 g
C26511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26515G Supplier TCI America Supplier No. C26515G

Chemical Identifiers

CAS 118289-55-7
Molecular Formula C10H9Cl2NO
Molecular Weight (g/mol) 230.088
MDL Number MFCD03411598
InChI Key ZTQQXEPZEYIVDK-UHFFFAOYSA-N
Synonym 6-Chloro-5-(2-chloroethyl)-2-indolinone
PubChem CID 10609474
IUPAC Name 6-chloro-5-(2-chloroethyl)-1,3-dihydroindol-2-one
SMILES C1C2=CC(=C(C=C2NC1=O)Cl)CCCl

Specifications

Melting Point 225°C
Quantity 5 g
UN Number 3077
Formula Weight 230.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-5-(2-chloroethyl)oxindole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.