Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-7-iodo-7-deazapurine 95.0+%, TCI America™
SDP

Catalog No. C31301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31301G 1 g
C31305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31301G Supplier TCI America Supplier No. C31301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123148-78-7
Molecular Formula C6H3ClIN3
Molecular Weight (g/mol) 279.465
MDL Number MFCD09263258
InChI Key CBWBJFJMNBPWAL-UHFFFAOYSA-N
Synonym 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine
PubChem CID 14809281
IUPAC Name 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=C(C2=C(N1)N=CN=C2Cl)I

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 1 g
Formula Weight 279.47
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.