Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-7-iodo-deazapurine, 96%

Catalog No. AC448850010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448850010 1 g
AC448850050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448850010 Supplier Thermo Scientific Chemicals Supplier No. 448850010
Add to Cart

CAS: 123148-78-7 | C6H3ClIN3 | 279.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123148-78-7
Molecular Formula C6H3ClIN3
Molecular Weight (g/mol) 279.47
InChI Key CBWBJFJMNBPWAL-UHFFFAOYSA-N
Synonym 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine
PubChem CID 14809281
IUPAC Name 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=C(C2=C(N1)N=CN=C2Cl)I

Specifications

Melting Point 179°C to 183°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 279.47
Percent Purity 96%
Chemical Name or Material 6-Chloro-7-iodo-deazapurine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.