Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-m-cresol 99.0+%, TCI America™
SDP

Catalog No. c07405g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Specifications

CAS 615-74-7
Melting Point 47°C
Color Red-White
Boiling Point 196°C
Molecular Formula C7H7ClO
Quantity 5 g
MDL Number MFCD00002175
UN Number 3437
Synonym 6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
InChI Key SMFHPCZZAAMJJO-UHFFFAOYSA-N
SMILES CC1=CC=C(Cl)C(O)=C1
IUPAC Name 2-chloro-5-methylphenol
Molecular Weight (g/mol) 142.58
PubChem CID 12008
Formula Weight 142.58
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-m-cresol
Show More Show Less
EINECSNumber (3)-0900
RTECSNumber SK4930000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.