Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™
SDP

Catalog No. C309725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C309725G 25 g
C30975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C309725G Supplier TCI America Supplier No. C309725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26518-76-3
Molecular Formula C10H8ClNO3
Molecular Weight (g/mol) 225.63
MDL Number MFCD01321312
InChI Key MIAHXWVABDHISZ-UHFFFAOYSA-N
PubChem CID 2764458
IUPAC Name 6-(2-chloroacetyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one
SMILES ClCC(=O)C1=CC=C2OCC(=O)NC2=C1

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 25 g
Formula Weight 225.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.