Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloroindole, 99%

Click to view available options
Quantity:
1g
5g
17422-33-2
C8H6ClN
151.593
MFCD00005681
YTYIMDRWPTUAHP-UHFFFAOYSA-N
6-chloroindole,1h-indole, 6-chloro,6-chloro indole,6-chloro-indole,6-chlor-1h-indole,pubchem1669,6-chloro-1-h-indole,#,ksc174m9p
87111
CHEBI:80918
6-chloro-1H-indole
C1=CC(=CC2=C1C=CN2)Cl
17422-33-2
C8H6ClN
151.593
MFCD00005681
YTYIMDRWPTUAHP-UHFFFAOYSA-N
6-chloroindole,1h-indole, 6-chloro,6-chloro indole,6-chloro-indole,6-chlor-1h-indole,pubchem1669,6-chloro-1-h-indole,#,ksc174m9p
87111
CHEBI:80918
6-chloro-1H-indole
C1=CC(=CC2=C1C=CN2)Cl
Melting Point 87°C to 90°C
Quantity 1g
Beilstein 112345
Formula Weight 151.6
Percent Purity 99%
Chemical Name or Material 6-Chloroindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning