Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloropyridazine-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AC429530050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 5096-73-1
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00160464
InChI Key HHGZQZULOHYEOH-UHFFFAOYSA-N
Synonym 6-chloro-pyridazine-3-carboxylic acid, 3-chloropyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 6-chloro, 6-chloro-3-carboxy-pyridazine, 3-carboxy-6-chloropyridazine, 6-chloro-3-pyridazine-carboxylic acid, 6-chloropyridazine-3-carboxylic, 6-chloropyridazine-3-carboxylicacid, pubchem17692
PubChem CID 6415762
IUPAC Name 6-chloropyridazine-3-carboxylic acid
SMILES OC(=O)C1=NN=C(Cl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 145.0°C to 148.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 158.54
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-chloropyridazine-3-carboxylic acid
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.