Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one 95.0+%, TCI America™
SDP

Catalog No. C24041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24041G 1 g
C2404200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24041G Supplier TCI America Supplier No. C24041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 128119-95-9
Molecular Formula C21H17N3O2
Molecular Weight (g/mol) 343.386
InChI Key DOIVGWIGBQTTHP-UHFFFAOYSA-N
PubChem CID 14512392
IUPAC Name 9-(diethylamino)-5-oxobenzo[a]phenoxazine-6-carbonitrile
SMILES CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C(=C3O2)C#N

Specifications

Quantity 1 g
Formula Weight 343.39
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.