Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 6 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 8 in. Sash Opening
SDP

Catalog No. 304681131 Shop All Labconco Corporation Products
Change view
Click to view available options
Plug Type:
China, Australia
India
Japan
Schuko
UK
Includes:
Accessory Pk.
Base Stand
Base Stand, Accessory Pk.
Biosafety Cabinet Only
Electrical Requirements:
100 V, 50/60 Hz, 12 A
230 V, 50/60 Hz, 6 A
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Includes Electrical Requirements
30-468-1131 Schuko Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-0021 Japan Biosafety Cabinet Only 100 V, 50/60 Hz, 12 A
30-468-0031 Schuko Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-468-0041 China, Australia Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-468-0051 UK Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-468-0071 India Biosafety Cabinet Only 230 V, 50/60 Hz, 6 A
30-468-0121 Japan Base Stand 100 V, 50/60 Hz, 12 A
30-468-0131 Schuko Base Stand 230 V, 50/60 Hz, 6 A
30-468-0141 China, Australia Base Stand 230 V, 50/60 Hz, 6 A
30-468-0151 UK Base Stand 230 V, 50/60 Hz, 6 A
30-468-0171 India Base Stand 230 V, 50/60 Hz, 6 A
30-468-1021 Japan Accessory Pk. 100 V, 50/60 Hz, 12 A
30-468-1031 Schuko Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1041 China, Australia Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1051 UK Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1071 India Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1121 Japan Base Stand, Accessory Pk. 100 V, 50/60 Hz, 12 A
30-468-1141 China, Australia Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1151 UK Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
30-468-1171 India Base Stand, Accessory Pk. 230 V, 50/60 Hz, 6 A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 30-468-1131 Supplier Labconco™ Supplier No. 304681131
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

6 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 8 in. Sash Opening is an unique, flexible BSC that can be operated as Type A or B and is the only Type C1 in the world.

  • Requires supporting base if included stand option is not selected
  • Intrinsically-safe negative pressure design includes:
    • Dual Electronically Commutated Motors (ECM)
    • Constant Airflow Profile™ (CAP) Technology airflow monitoring system
    • Active Protection Protocol operates the cabinet's exhaust blowers for for up to five minutes preventing the flow of air into the laboratory in the event of exhaust system failure
    • BSC required Air-Wave™ Entry System at access opening to enhance containment
    • Contain-Air™ Negative Pressure Channel at top of sash to enhance containment
    • Two programmable working sash heights (8 in. and 10 in.)
    • 10° angled, fully-closing, 1/4 in. tempered safety glass sash with counterbalanced, anti-racking, easy-to-lift mechanism
    • Curved waterfall design stainless steel inlet grille with Reserve-Air™ Secondary Airflow Slots
    • 22.6 in. (58 cm) maximum sash opening height and 27.0 in. (69 cm) viewing height Chem-Zone™ work surface with dedicated direct exhaust for use with hazardous vapors or radionuclides
    • Leak-tight interior with removable, type 304 stainless steel 3-piece work surface (dished Chem-Zone in center and two flat sides) with lift out knobs and clearly delineated working area
    • Nominal inflow velocity of 105 feet per minute (fpm) (0.53 m/sec)
    • Nominal downflow velocity of 65 fpm (0.33 m/sec) for 4 ft. models and 55 fpm (0.28 m/sec) for 6 ft. models
  • Touchpad on right-hand side post controls blower, light(s), timer, electrical receptacles, audible alarm mute and menu selection
  • Omni-Flex™ two mode design (Type A and B mode)
    • Low static pressure and volumetric rate exhaust requirements when in Type B mode
    • Night-Smart™ System for idling the blower when the sash is fully closed (while in Type A mode only)
  • MyLogic™ Operating System and interior-mounted, line-of-sight, full color, easy-to-understand LCD information center with:
    • Smart-Start™ System for start up and shut down operations
    • Filter Life Remaining bar graph for each HEPA filter
    • Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
    • Bright, 90-150 ft. candle, glare-free LED lighting located outside the contaminated work area
    • Interval/elapsed timer for experiment monitoring, LED light or UV light control (on models with UV light)
    • Electronic security lock (optional activation) that requires access code to operate the cabinet

Specifications

Description Class II Type C1 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA
Chamber Material Stainless Steel
Product Type Class II Biosafety Cabinet
Model Axiom
Noise Level 67 dBA
Outlets 2
Plug Type Schuko
Power Consumption 440 W
Receptacles 2
Sash Opening (Metric) 20 cm
Width (English) Exterior 78.3 in.
Volume (Metric) Exhaust 787 m3/hr.
Display Full-color LCD
Filter Efficiency 0.9999
Filter Type HEPA
Width (Metric) Exterior 198.9 cm
Housing Material Powder Coated Steel
Includes Base Stand, Accessory Pk.
Lighting LED
Sash Opening (English) 8 in.
Service Fixtures Two
Warranty 5 Years
Height (English) Exterior 64.2 in.
Height (Metric) Exterior 163.1 cm
Depth (English) Exterior 32.7 in.
Depth (Metric) Exterior 83.1 cm
Net Weight (English) 955 lb.
Shipping Weight (Metric) 533 kg
Electrical Requirements 230 V, 50/60 Hz, 6 A
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.