Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6 His Polyclonal Antibody, FITC, QED Bioscience™
SDP

Catalog No. 502277817
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7817 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7817 Supplier Leinco Technologies Inc Supplier No. 18812F100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

6 His Polyclonal antibody specifically detects 6 His in Tag samples. It is validated for Immunocytochemistry

Specifications

Antigen 6 His
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Goat
Immunogen 6-His (HHHHHH) conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.