Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6 His Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
$482.32 - $3247.60
Specifications
Antigen | 6 His |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7819
![]() ![]() |
Leinco Technologies Inc
1881401100UG |
100 μg |
Each for $482.32
|
|
|||||
50-227-7820
![]() ![]() |
Leinco Technologies Inc
188141MG |
1 mg |
Each for $3,247.60
|
|
|||||
Description
6 His Polyclonal antibody specifically detects 6 His in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
6 His | |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
PBS with 0.09% sodium azide; pH 7.2 | |
6-His (HHHHHH) conjugated to KLH. | |
Protein A | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
6His tag; 6X His; His Tag; Histidine Tag | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title