Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxycoumarin, 98%

Catalog No. AC459410050
Click to view available options
Quantity:
1 g
5 g

CAS: 6093-68-1 | C9H6O3 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6093-68-1
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.15
InChI Key CJIJXIFQYOPWTF-UHFFFAOYSA-N
Synonym 6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
PubChem CID 99477
IUPAC Name 6-hydroxychromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1O

Specifications

Melting Point 249.0°C to 253.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 6-Hydroxycoumarin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.