Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxydopamine Hydrobromide 99%, Thermo Scientific™

Catalog No. AC172250010
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-00-0
Molecular Formula C8H12BrNO3
Molecular Weight (g/mol) 250.092
InChI Key MLACDGUOKDOLGC-UHFFFAOYSA-N
Synonym 6-hydroxydopamine hydrobromide, oxidopamine hydrobromide, 6-hydroxydopamine hydrogen bromide, unii-twc1d4w0wb, 6-hydroxydopamine hbr, 5-2-aminoethyl benzene-1,2,4-triol hydrobromide, twc1d4w0wb, hydroxydopamine hydrobromide, 6-oh-da
PubChem CID 176170
IUPAC Name 5-(2-aminoethyl)benzene-1,2,4-triol;hydrobromide
SMILES C1=C(C(=CC(=C1O)O)O)CCN.Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Beige to Brown
Infrared Spectrum Authentic
Linear Formula (HO)3C6H2CH2CH2NH2.HBr
Quantity 1 g
Formula Weight 250.1
Percent Purity 98.5 to 101.5% (total base), ≥98.5% (TLC)
Physical Form Fine Crystalline Powder
Chemical Name or Material 6-Hydroxydopamine hydrobromide, 99%
EINECSNumber 211-247-0
RTECSNumber DC4600000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.