Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxyindazole 98.0+%, TCI America™
SDP

Catalog No. H13591G
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 23244-88-4
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00022788
InChI Key FVCQWGDUOULXIJ-UHFFFAOYSA-N
PubChem CID 90041
IUPAC Name 1,2-dihydroindazol-6-one
SMILES C1=CC2=CNNC2=CC1=O

Specifications

Melting Point 219°C
Quantity 1 g
Formula Weight 134.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxyindazole
RTECSNumber NK8010000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.