Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxyindole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2380-86-1
C8H7NO
133.15
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
6-hydroxyindole,6-indolol,indolol,6-hydroxy indole,unii-3i03jz599t,6-hydroxy-1h-indole,6hydroxyindole,6-hydroxy-indole,indol-6-ol,1h-indole-6-ol
524508
1H-indol-6-ol
C1=CC(=CC2=C1C=CN2)O
2380-86-1
C8H7NO
133.15
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
6-hydroxyindole,6-indolol,indolol,6-hydroxy indole,unii-3i03jz599t,6-hydroxy-1h-indole,6hydroxyindole,6-hydroxy-indole,indol-6-ol,1h-indole-6-ol
524508
1H-indol-6-ol
C1=CC(=CC2=C1C=CN2)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 1g
Melting Point 124.0°C to 130.0°C
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 133.15
Physical Form Shiny Crystalline Powder
Percent Purity 97%
Chemical Name or Material 6-Hydroxyindole

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti

GHS Signal Word: Danger

Recommended Storage : Product may darken during storage