Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxysalicylic acid, 98%, Thermo Scientific™

Catalog No. p-4228779
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 3147-64-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
InChI Key AAUQLHHARJUJEH-UHFFFAOYSA-N
Synonym 6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
PubChem CID 591524
IUPAC Name 2-hydroxy-6-methoxybenzoic acid
SMILES COC1=CC=CC(=C1C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 135.9°C to 136.4°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Formula Weight 168.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Methoxysalicylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.