Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-2-heptanone 98.0+%, TCI America™
SDP

Catalog No. M070025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M070025ML 25 mL
M07005ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M070025ML Supplier TCI America Supplier No. M070025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 928-68-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00026527
InChI Key DPLGXGDPPMLJHN-UHFFFAOYSA-N
Synonym Methyl 4-Methylpentyl Ketone
PubChem CID 13572
IUPAC Name 6-methylheptan-2-one
SMILES CC(C)CCCC(C)=O

Specifications

Color Yellow
Boiling Point 171°C
Quantity 25 mL
UN Number 1224
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Methyl-2-heptanone
EINECSNumber (2)-0542
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.