Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylchromone hydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
207511-19-1
C10H8O2·xH2O
160.17
MFCD00209598
QWIQUUMSUBFRAX-UHFFFAOYSA-N
6-methylchromone hydrate,6-methyl-4h-chromen-4-one hydrate,6-methylchromen-4-one hydrate,6-methyl-4h-1-benzopyran-4-one-water 1/1
16212708
6-methylchromen-4-one;hydrate
CC1=CC2=C(C=C1)OC=CC2=O.O
207511-19-1
C10H8O2·xH2O
160.17
MFCD00209598
QWIQUUMSUBFRAX-UHFFFAOYSA-N
6-methylchromone hydrate,6-methyl-4h-chromen-4-one hydrate,6-methylchromen-4-one hydrate,6-methyl-4h-1-benzopyran-4-one-water 1/1
16212708
6-methylchromen-4-one;hydrate
CC1=CC2=C(C=C1)OC=CC2=O.O
Melting Point 76.0°C to 78.0°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,5% naoh solution,,ethylacetate,alcohol
Formula Weight 160.17
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 6-Methylchromone hydrate
Show More Show Less