Learn More
6-Methylquinoline, 98%
Description
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.
Solubility
Not miscible or difficult to mix with water.
Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | -22°C |
| Density | 1.066 |
| Boiling Point | 257°C to 259°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.615 |
| Quantity | 5 g |
| Beilstein | 110336 |
| Sensitivity | Light sensitive |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 143.19 |
| Show More |
Safety and Handling
| Hazard Category | H302-H315 |
| Hazard Statement | GHS H Statement H301-H341-H315-H319 Toxic if swallowed. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
| EINECSNumber | 202-084-6 |
| RTECSNumber | VC0550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.