Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroindazole 98.0+%, TCI America™
SDP

Catalog No. N0399250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0399250G 250 g
N039925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0399250G Supplier TCI America Supplier No. N0399250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7597-18-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00005695
InChI Key ORZRMRUXSPNQQL-UHFFFAOYSA-N
Synonym 6-nitroindazole, 6-nitro-2h-indazole, 6-nitroisoindazole, 1h-indazole, 6-nitro, 2h-indazole, 6-nitro, indazole,6-nitro, ccris 3263, chembl54277, 6ni, 6-nitro-indazol
PubChem CID 24239
IUPAC Name 6-nitro-1H-indazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])NN=C2

Specifications

Melting Point 182°C
Quantity 250 g
Formula Weight 163.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitroindazole
RTECSNumber NK7962100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.