Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitropyridine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AAH3576603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3576603 1 g
AAH35766MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3576603 Supplier Thermo Scientific Chemicals Supplier No. H3576603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26893-68-5
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD04114205
InChI Key MIHFOFLNJIPDRY-UHFFFAOYSA-N
Synonym 6-nitropyridine-2-carboxylic acid, 6-nitropicolinic acid, 6-nitro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-nitro, picolinic acid, 6-nitro, 2-carboxy-6-nitropyridine, 2-pyridinecarboxylicacid, 6-nitro
PubChem CID 578841
SMILES C1=CC(=NC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
Formula Weight 168.11
Percent Purity 97%
Chemical Name or Material 6-Nitropyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.