Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6 Phosphofructo 2 Kinase Rabbit anti-Human, Mouse, Rat, Clone: 7X2H3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | 6 Phosphofructo 2 Kinase |
---|---|
Clone | 7X2H3 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
6 Phosphofructo 2 Kinase Monoclonal antibody specifically detects 6 Phosphofructo 2 Kinase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
6 Phosphofructo 2 Kinase | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
6PF-2-K/Fru-2,6-P2ase 3,6-bisphosphatase, 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase, 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3,6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, FLJ37326, inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, IPFK2, Renal carcinoma antigen NY-REN-56 | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human 6 Phosphofructo 2 Kinase (NP_004557.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7X2H3 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Endocrinology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5209 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title