Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Pyrrolidin-1-ylpyridine-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC38004DA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC38004DA 1 g
CC38004CB 250 mg
CC38004EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC38004DA Supplier Thermo Scientific Chemicals Supplier No. CC38004DA
Add to Cart

Chemical Identifiers

CAS 230618-24-3
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
MDL Number MFCD08435859
InChI Key SZOZYXCTOMKVTL-UHFFFAOYSA-N
PubChem CID 10821162
IUPAC Name 6-pyrrolidin-1-ylpyridine-2-carbaldehyde
SMILES O=CC1=NC(=CC=C1)N1CCCC1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 6-pyrrolidin-1-yl picolinaldehyde, 6-pyrrolidin-1-yl pyridine-2-carbaldehyde, 6-pyrrolidin-1-yl-picolinaldehyde, 6-pyrrolidinylpyridine-2-carbaldehyde, 6-1-pyrrolidinyl-2-pyridinecarboxaldehyde, 2-pyridinecarboxaldehyde,6-1-pyrrolidinyl, 2-pyridinecarboxaldehyde, 6-1-pyrrolidinyl
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.