Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Quinolinol 96.0+%, TCI America™
SDP

Catalog No. Q006025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q006025G 25 g
Q00605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q006025G Supplier TCI America Supplier No. Q006025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 580-16-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00047611
InChI Key OVYWMEWYEJLIER-UHFFFAOYSA-N
Synonym 6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline
PubChem CID 11374
ChEBI CHEBI:48994
IUPAC Name quinolin-6-ol
SMILES C1=CC2=C(C=CC(=C2)O)N=C1

Specifications

Quantity 25 g
Formula Weight 145.16
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Quinolinol
RTECSNumber VC4130000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.