Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-tert-Butyl-m-cresol, MP Biomedicals

Catalog No. ICN201929
Click to view available options
Quantity:
100 g

Chemical Identifiers

CAS 88-60-8
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
InChI Key XOUQAVYLRNOXDO-UHFFFAOYSA-N
Synonym 6-tert-butyl-m-cresol, 6-tert-butyl-3-methylphenol, 2-tert-butyl-5-methylphenol, m-cresol, 6-tert-butyl, 3-methyl-6-tert-butylphenol, phenol, 2-1,1-dimethylethyl-5-methyl, 5-methyl-2-tert-butylphenol, unii-uxv32mj2ca, 2-tert-butyl-5-methyl-phenol, 6-t-butyl-m-cresol
PubChem CID 6937
IUPAC Name 2-tert-butyl-5-methylphenol
SMILES CC1=CC(=C(C=C1)C(C)(C)C)O

Specifications

Quantity 100 g
Formula Weight 164.25
Chemical Name or Material 6-tert-Butyl-m-cresol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.