Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Thioguanine, 98%

Catalog No. aab2128006
Click to view available options
Quantity:
1 g
5 g

CAS: 154-42-7 | C5H5N5S | 167.19 g/mol

6-Thioguanine acts as an antineoplastic and purine antimetabolite. It is also useful as an inhibition of stimulated expression of TNF-related apoptosis-inducing ligand (TRAIL) protein. It is involved in the treatment of acute leukemias and Psoriasis. Further, it is used for the treatment of ulcerative colitis and autoimmune diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
PubChem CID 2723601
ChEBI CHEBI:9555
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

Melting Point >360°C
Odor Odorless
Assay Percent Range 0.98
Quantity 5 g
UN Number UN2811
Beilstein 157765
Merck Index 14,9337
Solubility Information Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.
IUPAC Name 2-amino-6,7-dihydro-3H-purine-6-thione
Formula Weight 167.19
Percent Purity 98%
Chemical Name or Material 6-Thioguanine
Show More Show Less
Hazard Category H301-H341
Hazard Statement GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-827-2
RTECSNumber UP0740000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.